missing translation for 'onlineSavingsMsg'
Learn More

Hach Lange™ Total Chlorine Reagent Powder Pillows

For total chlorine determination by the DPD method.

Brand:  Hach Lange™ 2105669


 View more versions of this product

Product Code. 10254042

  • £58.50 / Each
Estimated Shipment: 05-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2.

Pack of 100 powder pillows. Boric Acid free!
Specifications

Specifications

EPA compliant
All except DR/3000, DR/2000
100 Pack
DPD
10 mL
Total Chlorine, Bromine, Iodine
0.02 to 2.00 mg/L Cl2
Total Chlorine Reagent Powder Pillows
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DPD Total Chlorine Reagent

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Carboxylate salt(confidential), di-Sodium hydrogen phosphate, Potassium iodide,N,N-Diethylphenylend

SDS
Documents

Documents

Certificates
Special Offers

Special Offers