missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Total Chlorine Reagent Powder Pillows
For total chlorine determination by the DPD method.
£54.75
Product Code | Brand | Range | Volume (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Range | Volume (Metric) | For Use With (Equipment) | Price | Quantity & Availability | |||
10254042
|
Hach Lange™
2105669 |
0.02 to 2.00 mg/L Cl2 | 10 mL | All except DR/3000, DR/2000 |
£54.75
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||
13177429
|
Hach Lange™
1406499 |
0.02 to 2.00 mg/L Cl2 | 25 mL | DR/3000, DR/2000 |
£54.75
Pack of 100 |
Estimated Shipment: 28-10-2024
Log in to see stock. |
|||
Description
For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2.
Pack of 100 powder pillows. Boric Acid free!Specifications
100 Pack | |
Total Chlorine Reagent Powder Pillows |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- DPD Total Chlorine Reagent
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Carboxylate salt(confidential), di-Sodium hydrogen phosphate, Potassium iodide,N,N-Diethylphenylend