missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hach Lange™ Total Chlorine Reagent Powder Pillows
For total chlorine determination by the DPD method.
Brand: Hach Lange™ 1406499
This item is not returnable.
View return policy
Description
For use with Chlorine Visual Test Kits up to 3.5 mg/L Cl2.
Pack of 100 powder pillows.Boric Acid free!Specifications
EPA compliant | |
Total chlorine determination | |
Total Chlorine, Bromine, Iodine | |
0.02 to 2.00 mg/L Cl2 | |
Total Chlorine Reagent Powder Pillows |
25 mL | |
DR/3000, DR/2000 | |
100 Pack | |
8167 LR, DPD |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 14064-99 DPD Total Chlorine Reagent
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : di-Sodium hydrogen phosphate, Potassiumiodide, N,N-Diethylphenylendiammonium salt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction