Learn More
Masterflex™ Traceable™ Silicone Pump Tubing
Ideal for use with food, pharmaceuticals, and other critical solutions
Brand:  Masterflex™ 90024-26
 View more versions of this product
Product Code. 11761555
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Ideal for use with food, pharmaceuticals, and other critical solutions
  • Replacement tubing for Variable-Speed Pump
  • Can be sterilized by autoclaving
Specifications
Specifications
Autoclavable
1.58 mm
25 ft.
All Variable-Speed pumps
1/16 in.
Silicone
7.6 m
Pump Tubing