Learn More
Fisherbrand™ TraceableOne™ Single-Use USB Temperature Data Logger
The TraceableOne™ Single-Use USB Temperature Data Logger automatically generates PDF data report via USB port.
Brand:  Fisherbrand™ 18004-11
 View more versions of this product
Product Code. 17307629
  • £680.00 / Pack of 40
Explore more special offers
Description
Description
  • LED alarm indicator for visual status indication
  • Time delay of 30 minutes before logging begins for enhanced logging accuracy
  • Protective plastic pouch maintains waterproof IP67 rating
  • Bar code for in-house scanning
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
Specifications
Specifications
Single-use USB Temperature Datalogger
16,000 Readings, 90 Day, 10 Minute Interval
LED Indicator
2 Years
USB
-20°C to -15°C
No Display
NIST
3.0 V CR2450
1
Temperature