Learn More
Thermo Scientific™ TraceGOLD™ TG-624 and TG-624SilMS GC Columns
Optimize volatile organics analysis with this low-bleed, midpolarity GC column with high thermal stability.
£455.27 - £1507.50
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Stationary Phase | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Diameter (Metric) Inner | Film Thickness | Stationary Phase | Price | Quantity & Availability | ||||||
10066984
|
Thermo Scientific™
26085-3320 |
30 m | 0.25 mm | 1.4 μm | TraceGOLD™ TG-624 |
£618.08
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
15224523
|
Thermo Scientific™
26059-3390 |
30 m | 0.32 mm | 1.8 μm | TraceGOLD™ TG-624 SilMS |
£621.09
Each |
In Stock
Log in to see stock. |
||||||
15244523
|
Thermo Scientific™
26059-4950 |
20 m | 0.18 mm | 1 μm | TraceGOLD™ TG-624 SilMS |
£455.27
Each |
In Stock
Log in to see stock. |
||||||
15274523
|
Thermo Scientific™
26059-4080 |
60 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 SilMS |
£1030.13
Each |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
15294523
|
Thermo Scientific™
26059-4900 |
75 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 SilMS | N/A | N/A | N/A | |||||
15204533
|
Thermo Scientific™
26059-4090 |
105 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 SilMS | N/A | N/A | N/A | |||||
15284523
|
Thermo Scientific™
26059-3410 |
60 m | 0.32 mm | 1.8 μm | TraceGOLD™ TG-624 SilMS |
£989.93
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
10566575
|
Thermo Scientific™
26085-3330 |
60 m | 0.25 mm | 1.4 μm | TraceGOLD™ TG-624 |
£994.95
Each |
Estimated Shipment: 14-05-2025
Log in to see stock. |
||||||
10669875
|
Thermo Scientific™
26085-3390 |
30 m | 0.32 mm | 1.8 μm | TraceGOLD™ TG-624 |
£668.33
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
10240055
|
Thermo Scientific™
26085-3410 |
60 m | 0.32 mm | 1.8 μm | TraceGOLD™ TG-624 |
£1095.45
Each |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
10197844
|
Thermo Scientific™
26085-3960 |
30 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 |
£713.55
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
10753307
|
Thermo Scientific™
26085-4080 |
60 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 |
£1135.65
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
10016074
|
Thermo Scientific™
26085-4090 |
105 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 |
£1507.50
Each |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
10535255
|
Thermo Scientific™
26085-4900 |
75 m | 0.53 mm | 3 μm | TraceGOLD™ TG-624 |
£1412.03
Each |
Estimated Shipment: 21-05-2025
Log in to see stock. |
||||||
10280385
|
Thermo Scientific™
26085-4950 |
20 m | 0.18 mm | 1 μm | TraceGOLD™ TG-624 |
£512.55
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
10763307
|
Thermo Scientific™
26085-4960 |
40 m | 0.18 mm | 1 μm | TraceGOLD™ TG-624 |
£899.48
Each |
Estimated Shipment: 19-05-2025
Log in to see stock. |
||||||
Beschreibung
Analyze residual solvents, volatile organic compounds, alcohols, and oxygenates using Thermo Scientific™ TraceGOLD™ TG-624 and TG-624SilMS GC Columns. Both phases offer the low to mid polarity phase, 6% cyanopropylphenyl methypolysilozane.
- TraceGOLD TG-624 GC Columns provide 90+% resolution of the first six gases in EPA Method 8260 and EPA Method 524.4. These columns are also ideal for EPA Methods 624 and 608.
- TraceGOLD TG-624SilMS GC Columns provide high thermal stability with maximum temperatures up to 320°C, which is ideal for volatile organics analysis. They are highly inert, providing excellent peak shape for a wide range of compounds.
- Residual solvents
- Volatile organic compounds
- Alcohols
- Oxygenates
Spezifikation
G43 | |
Mid-Polar |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.