Learn More
trans-2,4-Dichlorocinnamic acid, 98%, Thermo Scientific™
£19.50 - £209.00
Chemical Identifiers
CAS | 20595-45-3 |
---|---|
Molecular Formula | C9H6Cl2O2 |
Molecular Weight (g/mol) | 217.045 |
MDL Number | MFCD00004373 |
InChI Key | MEBWABJHRAYGFW-DUXPYHPUSA-N |
Synonym | 2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid |
PubChem CID | 688026 |
IUPAC Name | (E)-3-(2,4-dichlorophenyl)prop-2-enoic acid |
SMILES | C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
20595-45-3 | |
217.045 | |
MEBWABJHRAYGFW-DUXPYHPUSA-N | |
688026 | |
C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O |
C9H6Cl2O2 | |
MFCD00004373 | |
2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid | |
(E)-3-(2,4-dichlorophenyl)prop-2-enoic acid |
Specifications
20595-45-3 | |
C9H6Cl2O2 | |
1369152 | |
MEBWABJHRAYGFW-DUXPYHPUSA-N | |
(E)-3-(2,4-dichlorophenyl)prop-2-enoic acid | |
688026 | |
98% |
233°C to 235°C | |
MFCD00004373 | |
2,4-dichlorocinnamic acid, trans-2,4-dichlorocinnamic acid, e-3-2,4-dichlorophenyl acrylic acid, cinnamic acid, 2,4-dichloro, 3-2,4-dichlorophenyl-2-propenoic acid, unii-c09kk5d2t2, 2-propenoic acid, 3-2,4-dichlorophenyl-, 2e, 2e-3-2,4-dichlorophenyl acrylic acid, 2-propenoic acid, 3-2,4-dichlorophenyl, 2e-3-2,4-dichlorophenyl prop-2-enoic acid | |
C1=CC(=C(C=C1Cl)Cl)C=CC(=O)O | |
217.045 | |
217.05 | |
trans-2,4-Dichlorocinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'rtecsNumber' : GD8575000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures