missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-3-(Boc-amino)cyclobutanecarboxylic acid, 97%, Thermo Scientific Chemicals
£72.20 - £168.00
Chemical Identifiers
CAS | 939400-34-7 |
---|---|
Molecular Formula | C10H17NO4 |
Molecular Weight (g/mol) | 215.249 |
MDL Number | MFCD06657680 |
InChI Key | KLCYDBAYYYVNFM-UHFFFAOYSA-N |
Synonym | cis-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, 3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclobutane-1-carboxylic acid, cis-3-boc-aminocyclobutanecarboxylic acid |
PubChem CID | 24720963 |
IUPAC Name | 3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid |
SMILES | CC(C)(C)OC(=O)NC1CC(C1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15447976
|
Thermo Scientific Alfa Aesar
H62902.MD |
250 mg |
£72.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15419358
|
Thermo Scientific Alfa Aesar
H62902.03 |
1 g |
£168.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
939400-34-7 | |
215.249 | |
KLCYDBAYYYVNFM-UHFFFAOYSA-N | |
24720963 | |
CC(C)(C)OC(=O)NC1CC(C1)C(=O)O |
C10H17NO4 | |
MFCD06657680 | |
cis-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, 3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclobutane-1-carboxylic acid, cis-3-boc-aminocyclobutanecarboxylic acid | |
3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid |
Specifications
939400-34-7 | |
MFCD06657680 | |
KLCYDBAYYYVNFM-UHFFFAOYSA-N | |
3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclobutane-1-carboxylic acid | |
24720963 | |
97% |
C10H17NO4 | |
cis-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, 3-tert-butoxycarbonylamino cyclobutanecarboxylic acid, trans-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, 3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-tert-butoxycarbonyl amino cyclobutanecarboxylic acid, cis-3-boc-amino cyclobutanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclobutane-1-carboxylic acid, cis-3-boc-aminocyclobutanecarboxylic acid | |
CC(C)(C)OC(=O)NC1CC(C1)C(=O)O | |
215.249 | |
215.24 | |
trans-3-(Boc-amino)cyclobutanecarboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only