missing translation for 'onlineSavingsMsg'
Learn More

trans-3-Octene, 97%, Thermo Scientific Chemicals

£95.64 - £381.00

Chemical Identifiers

CAS 14919-01-8
Molecular Formula C8H16
Molecular Weight (g/mol) 112.216
MDL Number MFCD00039971
InChI Key YCTDZYMMFQCTEO-FNORWQNLSA-N
Synonym trans-3-octene, e-3-octene, e-oct-3-ene, 3-octene, e, 3-octene, 3e, unii-2a4p9ko860, 3-octene, 3e-3-octene, 3e-oct-3-ene, cis-3-octene
PubChem CID 638228
IUPAC Name (E)-oct-3-ene
SMILES CCCCC=CCC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304838
SDS View Documents
Thermo Scientific Alfa Aesar
043154.09
10 g N/A N/A N/A
11314838
SDS View Documents
Thermo Scientific Alfa Aesar
043154.18
50 g N/A N/A N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225
Highly flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 238-990-3

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
trans-3-Octene, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.