Learn More
trans-4-Methoxycinnamic acid, 98%, Thermo Scientific Chemicals
Description
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Esters derived from trans-4-methoxycinnamic acid are effective absorbers of UV radiation. The starting material was trans-4-methoxycinnamic acid and 3,5dimethoxybenzoic acid methyl ester in isolated avenanthramide alkaloids synthsis. trans-4-Methoxycinnamic acid was converted into its acid chloride (yield 98%) with thionyl chloride. Employed in organic synthesis of pharmaceutical intermediates, synthetic anti-adrenergic drugs esmolol, cosmetics ultraviolet absorption.
Solubility
Soluble in ethanol, ethyl acetate and other organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 171°C to 175°C |
Quantity | 10 g |
Beilstein | 510468 |
Solubility Information | Soluble in ethanol,ethyl acetate and other organic solvents. |
Formula Weight | 178.19 |
Percent Purity | 98% |
Chemical Name or Material | trans-4-Methoxycinnamic acid |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10137632 | Certificate of Analysis | 22/11/2021 | A13870.18,A13870.30,A13870.09 |
10179420 | Certificate of Analysis | 22/11/2021 | A13870.18,A13870.30,A13870.09 |
10034166 | Certificate of Analysis | 22/11/2021 | A13870.18,A13870.30,A13870.09 |
10194202 | Certificate of Analysis | 22/11/2021 | A13870.18,A13870.30,A13870.09 |
10234949 | Certificate of Analysis | 22/11/2021 | A13870.18,A13870.30,A13870.09 |
Safety and Handling

- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : trans-4-Methoxycinnamic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.