missing translation for 'onlineSavingsMsg'
Learn More

trans-4-tert-Pentylcyclohexanol, 97%, Thermo Scientific™

£28.00 - £87.10

Chemical Identifiers

CAS 20698-30-0
Molecular Formula C11H22O
Molecular Weight (g/mol) 170.30
MDL Number MFCD00067157,MFCD00019356
InChI Key VDQZABQVXYELSI-UHFFFAOYSA-N
Synonym 4-tert-amylcyclohexanol, 4-tert-pentyl cyclohexanol, trans-4-tert-pentyl cyclohexanol, p-tert-amylcyclohexanol, trans-4-tert-pentylcyclohexanol, cyclohexanol, 4-tert-pentyl, cyclohexanol, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl cyclohexan-1-ol, 4-tert-pentylcyclohexanol, 4-2-methylbutan-2-yl cyclohexanol
PubChem CID 79302
IUPAC Name 4-(2-methylbutan-2-yl)cyclohexan-1-ol
SMILES CCC(C)(C)C1CCC(O)CC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11427014
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21479.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11437014
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B21479.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

20698-30-0
170.30
VDQZABQVXYELSI-UHFFFAOYSA-N
79302
CCC(C)(C)C1CCC(O)CC1
C11H22O
MFCD00067157,MFCD00019356
4-tert-amylcyclohexanol, 4-tert-pentyl cyclohexanol, trans-4-tert-pentyl cyclohexanol, p-tert-amylcyclohexanol, trans-4-tert-pentylcyclohexanol, cyclohexanol, 4-tert-pentyl, cyclohexanol, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl cyclohexan-1-ol, 4-tert-pentylcyclohexanol, 4-2-methylbutan-2-yl cyclohexanol
4-(2-methylbutan-2-yl)cyclohexan-1-ol

Specifications

20698-30-0
154°C to 156°C (40 mmHg)
MFCD00067157,MFCD00019356
VDQZABQVXYELSI-UHFFFAOYSA-N
4-(2-methylbutan-2-yl)cyclohexan-1-ol
79302
97%
24°C to 27°C
C11H22O
4-tert-amylcyclohexanol, 4-tert-pentyl cyclohexanol, trans-4-tert-pentyl cyclohexanol, p-tert-amylcyclohexanol, trans-4-tert-pentylcyclohexanol, cyclohexanol, 4-tert-pentyl, cyclohexanol, 4-1,1-dimethylpropyl, 4-2-methylbutan-2-yl cyclohexan-1-ol, 4-tert-pentylcyclohexanol, 4-2-methylbutan-2-yl cyclohexanol
CCC(C)(C)C1CCC(O)CC1
170.30
170.3
trans-4-tert-Pentylcyclohexanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications