Learn More
Brand™ Multi-Channel Micropipette Transferpette™ -12 Electronic, DE-M
Ergonomic, handy design. Clear and intuitive operation. Up to 4,000 pipetting cycles on a single charge.
Brand: Brand™ 705463
693.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Transferpette electronic pipettes combine the globally proven properties of mechanical piston operated air displacement pipettes by BRAND with the advantages of an electronic device.
- Features: The pipette shaft can be unscrewed and is entirely autoclavable (121 °C).
- Functions: pipetting, reverse pipetting, mixing, dispensing, up to 300 μl also GEL-Electrophoresis.
Easy Calibration-Technique for making quick adjustments of the Transferpette™ electronic, without tools. - Limitations of use: Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F) (other temperatures upon request)
- Vapor pressure up to 500 mbar
- Viscosity: 260 mPa s
- For viscous media, the speed must be adjusted if necessary.

Specifications
≤ 0.8%, ± 0.8μL | |
CE-Marked | |
Yellow | |
Transferpette | |
Electronic Microliter Pipette | |
LCD | |
UK/Ireland AC Adapter | |
1 Each | |
230 V |
360° | |
Autoclavable | |
12 | |
Yes | |
5 to 100 μL | |
Ergonomic | |
0.1μL | |
230V/50 Hz | |
50 Hz |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.