Learn More
VITLAB™ VITsafe™ 1000mL Wide-Mouth Wash Bottles
Imprinted with durable safety symbols, and other important information for optimum laboratory safety. Vitlab™ VITsafe™ Wide-Mouth Wash Bottles include color coded vent-cap that supports easy visual identification of the media.
Brand: VITLAB™ 1453829
Additional Details : Weight : 0.12000kg
Description
- Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
- Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
- Imprinted with:
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
- The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping
Specifications
1000 mL | |
Polypropylene | |
Directive (EC) No. 1272/2008 (GHS) | |
Wide | |
Polypropylene | |
Vented |
Imprint: Acetone | |
Red | |
Yes | |
226 mm | |
91 mm |