missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ 1000mL Wide-Mouth Wash Bottles
Imprinted with durable safety symbols, and other important information for optimum laboratory safety. Vitlab™ VITsafe™ Wide-Mouth Wash Bottles include color coded vent-cap that supports easy visual identification of the media.
£11.80 - £139.00
Specifications
Capacity (Metric) | 1000 mL |
---|---|
Closure Color | Red |
Certifications/Compliance | Directive (EC) No. 1272/2008 (GHS) |
Closure Included | Yes |
Mouth | Wide |
Product Code | Brand | Material | Description | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Description | Price | Quantity & Availability | ||||
15762066
|
VITLAB™
1353819 |
LDPE | Imprint: Distilled Water |
£139.00
Pack of 12 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15772066
|
VITLAB™
1353839 |
LDPE | Imprinted: Methanol |
£139.00
Pack of 12 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15782066
|
VITLAB™
1353849 |
LDPE | Imprinted: Isopropanol |
£139.00
Pack of 12 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
15792066
|
VITLAB™
1353859 |
LDPE | Imprinted: Ethyl Acetate |
£139.00
Pack of 12 |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||||
15702076
|
VITLAB™
1353869 |
LDPE | Imprinted: Ethanol |
£139.00
Pack of 12 |
Estimated Shipment: 15-07-2024
Log in to see stock. |
||||
15742076
|
VITLAB™
1453829 |
Polypropylene | Imprint: Acetone |
£138.00
Pack of 12 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||||
10287802
|
VITLAB™
1453829 |
Polypropylene | Imprint: Acetone |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
- Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
- Imprinted with:
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
- The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping
Specifications
1000 mL | |
Directive (EC) No. 1272/2008 (GHS) | |
Wide | |
Polypropylene | |
Vented |
Red | |
Yes | |
226 mm | |
91 mm |