missing translation for 'onlineSavingsMsg'
Learn More

VITLAB™ VITsafe™ 1000mL Wide-Mouth Wash Bottles

Imprinted with durable safety symbols, and other important information for optimum laboratory safety. Vitlab™ VITsafe™ Wide-Mouth Wash Bottles include color coded vent-cap that supports easy visual identification of the media.

Brand:  VITLAB™ 1353869

Additional Details : Weight : 1.82700kg

 View more versions of this product

Product Code. 15702076

  • £139.00 / Pack of 12
Estimated Shipment: 08-08-2024
to see stock.
Add to basket

Description

Description

  • Manufactured from PE-LD (or PP for Acetone and MEK), spray tube from PP
  • Complies in accordance to Directive (EC) No. 1272/2008 (GHS)
  • Imprinted with:
    • Material name in German, English, French and Spanish
    • Chemical formula, CAS No, hazard pictogram, signal word
    • Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
  • Precise spray jet and optimised medium backflow through the continuous and finely drawn tip of the spray tube
  • The VENT-CAP design oprevents almost all static overpressure minimizing leakage or dripping
Specifications

Specifications

1000 mL
LDPE
Directive (EC) No. 1272/2008 (GHS)
Wide
Polypropylene
Vented
Imprinted: Ethanol
Red
Yes
226 mm
91 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers