Learn More
BRAND™ Blaubrand™ Eterna™ AR-GLAS™ Type 3 Class AS Measuring Pipettes
Manufactured from AR-GLAS™ Soda lime glass, this type 3 Class AS Measuring Pipet includes graduations with DE-M marking and is calibrated for total delivery.
Brand:  BRAND™ 27319
 View more versions of this product
Product Code. 10603561
  • £87.85 / Pack of 12
In Stock
to see stock.
Explore more special offers
Add to basket
Beschreibung
Beschreibung
  • Manufactured from AR-GLAS™ Soda lime glass
  • Type 3 (zero on top)
  • DE-M Marking graduations
  • Calibrated to deliver (TD, Ex.)
  • Supplied with batch number and one batch certificate per packing unit
  • USP individual certificate or DAkkS calibration certificate available upon request
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
DIN EN ISO 835
Yes
±0.070 mL
AR-GLAS
0.02 mL
2 mL
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Sonderangebote
Sonderangebote