Learn More
VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles
Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks. Constructed of chemically resistant material for use with acetone and MEK.
Brand: VITLAB™ 1432989
Description
Working with chemical substances that can be dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.Constructed of durable, transparent polypropylene
- PP adds a significantly higher level of resistance to many solvents, including acetone and MEK
- Wash bottles and spray tubes made from PP may be autoclaved at 121°C for 20 minutes
- Transparent material allows for easy identification and monitoring
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- In accordance to EC Directive No. 1272/2008 (GHS)
- Design eliminates almost all static overpressure
- Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
- Color-coded caps support easy visual identification of the media
- Bottles utilize screw cap lids with GL threads for a strong, secure hold
- Finely drawn tip creates a precise spray jet for fine control
- Optimized medium backflow through the continuous spray tube
Specifications
| 500 mL | |
| Polypropylene | |
| 6Pack | |
| GL 25 | |
| 180 mm | |
| Methyl Ethyl Ketone | |
| Vented Screw Cap |
| Transparent | |
| Individually wrapped in PE bag | |
| Red | |
| Narrow | |
| Safety Wash Bottle | |
| 74 mm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.