missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles
Work safely with potentially dangerous chemicals. VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles feature a permanent safety imprint, color-coded VENT-CAP and precise spray control to eliminate drips and leaks. Constructed of chemically resistant material for use with acetone and MEK.
£80.45
Specifications
Color | Transparent |
---|---|
Material | Polypropylene |
Packaging | Individually wrapped in PE bag |
Screw Color | Red |
Thread Size (Metric) | GL 25 |
Description
Working with chemical substances that can be dangerous requires a high level of responsibility and care. VITsafe™ safety wash-bottles fulfill all safety requirements for safe handling. Spray bottles are individually wrapped in clearly labeled polyethylene bags.Constructed of durable, transparent polypropylene
- PP adds a significantly higher level of resistance to many solvents, including acetone and MEK
- Wash bottles and spray tubes made from PP may be autoclaved at 121°C for 20 minutes
- Transparent material allows for easy identification and monitoring
- Material name in German, English, French and Spanish
- Chemical formula, CAS No, hazard pictogram, signal word
- Risk Phrases (H phrases), safety phrases (P phrases), as well as NFPA Code
- In accordance to EC Directive No. 1272/2008 (GHS)
- Design eliminates almost all static overpressure
- Prevents virtually all leaking or dripping that can occur with temperature changes in the lab
- Color-coded caps support easy visual identification of the media
- Bottles utilize screw cap lids with GL threads for a strong, secure hold
- Finely drawn tip creates a precise spray jet for fine control
- Optimized medium backflow through the continuous spray tube
Specifications
Transparent | |
Individually wrapped in PE bag | |
GL 25 | |
Safety Wash Bottle |
Polypropylene | |
Red | |
Narrow | |
Vented Screw Cap |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
VITLAB™ VITsafe™ Narrow Mouth PP Safety Wash Bottles