Learn More
VITLAB™ Wash Bottles, Transparent Polypropylene
Wash equipment and materials with a precise spray jet and optimized backflow. VITLAB™ Wash Bottles, Transparent Polypropylene feature a screw cap with an incorporated spray tube on a narrow- or wide-mouth bottle. Spray bottles are individually wrapped, autoclavable and recyclable.
Brand: VITLAB™ 93793
Additional Details : Weight : 4.02000kg
Description
Wash bottles are appropriate for cleaning, coating and dousing in any laboratory environment. Polypropylene bottles are ideal for a variety of applications, including use with a range of chemicals, solvents and temperatures. Spray bottles are individually wrapped in clearly labeled polyethylene bags.- Designed for precise spray control
- Finely drawn tip creates a precise spray jet for fine control
- Optimised medium backflow through the continuous spray tube
- Choice of narrow- or wide-mouth bottles
- Narrow-mouth, tapered bottles for easier handling
- Wide-mouth, square-shouldered bottles for easy and safe filling
- All bottles feature screw cap lids with GL threads for a strong, secure hold
- Constructed of durable, transparent polypropylene
- Chemically resistant for use with liquid or powdered reagents
- Temperature resistant and autoclavable
- Transparent material for easy identification and monitoring
Specifications
250 mL | |
Polypropylene | |
GL 45 | |
Wide | |
Wash Bottle | |
Screw Cap |
Transparent | |
12Pack | |
Autoclavable | |
146 mm | |
58 mm |