missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VITLAB™ Wash Bottles, Transparent Polypropylene
Wash equipment and materials with a precise spray jet and optimized backflow. VITLAB™ Wash Bottles, Transparent Polypropylene feature a screw cap with an incorporated spray tube on a narrow- or wide-mouth bottle. Spray bottles are individually wrapped, autoclavable and recyclable.
£54.35 - £114.00
Specifications
Color | Transparent |
---|---|
Material | Polypropylene |
Quantity | 12Pack |
Autoclavable | Autoclavable |
Product Type | Wash Bottle |
Product Code | Brand | Capacity (Metric) | Thread Size (Metric) | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Thread Size (Metric) | Height (Metric) | Price | Quantity & Availability | |||
15712026
|
VITLAB™
93793 |
250 mL | GL 45 | 146 mm |
£63.80
Pack of 12 |
Estimated Shipment: 01-08-2024
Log in to see stock. |
|||
15742026
|
VITLAB™
94993 |
250 mL | GL 25 | 135 mm |
£54.35
Pack of 12 |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||
15722026
|
VITLAB™
93993 |
500 mL | GL 45 | 166 mm |
£84.45
Pack of 12 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
|||
15732026
|
VITLAB™
94193 |
1000 mL | GL 63 | 226 mm |
£114.00
Pack of 12 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||
15752026
|
VITLAB™
95193 |
1000 mL | GL 32 | 215 mm |
£94.20
Pack of 12 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||
Description
Wash bottles are appropriate for cleaning, coating and dousing in any laboratory environment. Polypropylene bottles are ideal for a variety of applications, including use with a range of chemicals, solvents and temperatures. Spray bottles are individually wrapped in clearly labeled polyethylene bags.- Designed for precise spray control
- Finely drawn tip creates a precise spray jet for fine control
- Optimised medium backflow through the continuous spray tube
- Choice of narrow- or wide-mouth bottles
- Narrow-mouth, tapered bottles for easier handling
- Wide-mouth, square-shouldered bottles for easy and safe filling
- All bottles feature screw cap lids with GL threads for a strong, secure hold
- Constructed of durable, transparent polypropylene
- Chemically resistant for use with liquid or powdered reagents
- Temperature resistant and autoclavable
- Transparent material for easy identification and monitoring
Specifications
Transparent | |
12Pack | |
Wash Bottle |
Polypropylene | |
Autoclavable | |
Screw Cap |