missing translation for 'onlineSavingsMsg'
Learn More

JULABO™ Standard Carrier Tray for Erlenmeyer Flasks

For use with Erlenmeyer flasks. Suitable for JULABO™ SW22 / SW23.

Brand:  JULABO™ 8970366

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10710651

  • £359.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Add to basket

Description

Description

  • Metal Construction
  • For use with JULABO™ SW22/SW23
  • Several sizes available
  • Choose a model to hold as few as 5 x 1000 mL Erlenmeyer Flasks or as many as 45 x 25 mL
Specifications

Specifications

SW22, SW23 shaking water bath
Carrier Tray
5 Erlenmeyer Flasks, 1000 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers