Learn More
Tri(1-aziridinyl)phosphine sulfide, 98%, Thermo Scientific™
£112.00 - £432.00
Chemical Identifiers
CAS | 52-24-4 |
---|---|
Molecular Formula | C6H12N3PS |
Molecular Weight (g/mol) | 189.22 |
MDL Number | MFCD00145452 |
InChI Key | FOCVUCIESVLUNU-UHFFFAOYSA-N |
Synonym | thiotepa, thio-tepa, triethylenethiophosphoramide, thiophosphamide, tiofosfamid, tiofosyl, thiofozil, tespamin, thioplex, tiofozil |
PubChem CID | 5453 |
ChEBI | CHEBI:9570 |
IUPAC Name | tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane |
SMILES | S=P(N1CC1)(N1CC1)N1CC1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in benzene, acetone and methanol.
Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
Chemical Identifiers
52-24-4 | |
189.22 | |
FOCVUCIESVLUNU-UHFFFAOYSA-N | |
5453 | |
tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane |
C6H12N3PS | |
MFCD00145452 | |
thiotepa, thio-tepa, triethylenethiophosphoramide, thiophosphamide, tiofosfamid, tiofosyl, thiofozil, tespamin, thioplex, tiofozil | |
CHEBI:9570 | |
S=P(N1CC1)(N1CC1)N1CC1 |
Specifications
52-24-4 | |
MFCD00145452 | |
thiotepa, thio-tepa, triethylenethiophosphoramide, thiophosphamide, tiofosfamid, tiofosyl, thiofozil, tespamin, thioplex, tiofozil | |
FOCVUCIESVLUNU-UHFFFAOYSA-N | |
tris(aziridin-1-yl)-sulfanylidene-$l^{5}-phosphane | |
5453 | |
189.22 | |
Solid |
C6H12N3PS | |
UN2811 | |
Soluble in benzene,acetone and methanol. | |
S=P(N1CC1)(N1CC1)N1CC1 | |
189.22 | |
CHEBI:9570 | |
98% | |
Tri(1-aziridinyl)phosphine sulfide |
Safety and Handling
GHS H Statement
H300-H340-H350
Fatal if swallowed.
May cause genetic defects.
May cause cancer.
P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
H300-H340-H350-H500
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 200-135-7
missing translation for 'rtecsNumber' : SZ2975000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold