Learn More
Tri-n-butyl(1-propenyl)tin, cis + trans, 94%, Thermo Scientific Chemicals
£36.07 - £302.64
Chemical Identifiers
CAS | 105494-65-3 |
---|---|
Molecular Formula | C15H32Sn |
Molecular Weight (g/mol) | 331.131 |
MDL Number | MFCD03425868 |
InChI Key | FJXYMVBFBYAWDR-UHFFFAOYSA-N |
Synonym | stannane, tributyl-1-propenyl, z-tri-n-butyl 1-propenyl tin, tri-n-butyl 1-propenyl tin, cis + trans, tributyl prop-1-en-1-yl stannane |
PubChem CID | 57369809 |
IUPAC Name | tributyl(prop-1-enyl)stannane |
SMILES | CCCC[Sn](CCCC)(CCCC)C=CC |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105494-65-3 | |
331.131 | |
FJXYMVBFBYAWDR-UHFFFAOYSA-N | |
57369809 | |
CCCC[Sn](CCCC)(CCCC)C=CC |
C15H32Sn | |
MFCD03425868 | |
stannane, tributyl-1-propenyl, z-tri-n-butyl 1-propenyl tin, tri-n-butyl 1-propenyl tin, cis + trans, tributyl prop-1-en-1-yl stannane | |
tributyl(prop-1-enyl)stannane |
Specifications
105494-65-3 | |
C15H32Sn | |
UN2788 | |
stannane, tributyl-1-propenyl, z-tri-n-butyl 1-propenyl tin, tri-n-butyl 1-propenyl tin, cis + trans, tributyl prop-1-en-1-yl stannane | |
CCCC[Sn](CCCC)(CCCC)C=CC | |
331.131 | |
331.13 | |
Tri-n-butyl(1-propenyl)tin, Cis + Trans |
77°C to 78°C (0.25 mmHg) | |
MFCD03425868 | |
4176362 | |
FJXYMVBFBYAWDR-UHFFFAOYSA-N | |
tributyl(prop-1-enyl)stannane | |
57369809 | |
94% |
Safety and Handling
GHS H Statement
H301-H372-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P260-P280-P301+P310-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
Recommended Storage : 2° to 8°C
RUO – Research Use Only