missing translation for 'onlineSavingsMsg'
Learn More
Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals Product Code.: 10072670

Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals

Product Code. 10072670
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10072670

Brand: Thermo Scientific Acros 207600250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.64
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209pegShow MoreShow Less
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 50.0°C to 55.0°C
Color White to Yellow
Boiling Point 201.0°C to 202.0°C (2.0 mmHg)
Flash Point 230°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (including isomer) (GC)
Packaging Glass bottle
Linear Formula [CH3(CH2)7]3P(O)
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in cyclohexane,methanol
Formula Weight 386.64
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467319 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0466187 Certificate TypeCertificate of Analysis Date16/09/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0463093 Certificate TypeCertificate of Analysis Date27/05/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0461145 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number207605000,207600250,207601000
Lot NumberA0444532 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog Number207605000,207600250,207601000
compliance-icons compliance-icons
Product Identifier
  • Tri-n-octylphosphine oxide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tri-n-octylphosphine oxide, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.