Learn More
Tributyl phosphate, 99+%, Thermo Scientific Chemicals
£44.60 - £302.00
Chemical Identifiers
CAS | 126-73-8 |
---|---|
Molecular Formula | C12H27O4P |
Molecular Weight (g/mol) | 266.32 |
MDL Number | MFCD00009436 |
InChI Key | STCOOQWBFONSKY-UHFFFAOYSA-N |
Synonym | tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de |
PubChem CID | 31357 |
ChEBI | CHEBI:35019 |
IUPAC Name | tributyl phosphate |
SMILES | CCCCOP(=O)(OCCCC)OCCCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10173990
|
Thermo Scientific Acros
169930010 |
1 L | Glass Bottle |
£44.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10337540
|
Thermo Scientific Acros
169930025 |
2.5 L | Glass Bottle |
£101.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11975141
|
Thermo Scientific Acros
169930100 |
10 L | Metal drum |
£302.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
126-73-8 | |
266.32 | |
STCOOQWBFONSKY-UHFFFAOYSA-N | |
31357 | |
tributyl phosphate |
C12H27O4P | |
MFCD00009436 | |
tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de | |
CHEBI:35019 | |
CCCCOP(=O)(OCCCC)OCCCC |
Specifications
126-73-8 | |
0.9790g/mL | |
146°C | |
99% min. (GC) | |
[CH3(CH2)3O]3P(O) | |
MFCD00009436 | |
15,9781 | |
tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de | |
STCOOQWBFONSKY-UHFFFAOYSA-N | |
tributyl phosphate | |
31357 | |
3.5 mPa.s (25°C) | |
99+% | |
Tributyl phosphate, 99% |
-79.0°C | |
289.0°C | |
Authentic | |
C12H27O4P | |
1.4230 to 1.4250 | |
01,II,397 | |
0.979 | |
Solubility in water: 0.6g/100mL (20°C). Other solubilities: soluble in alcohols,ketones,ethers,benzene,,carbon disulfide and most organic solvents | |
CCCCOP(=O)(OCCCC)OCCCC | |
266.32 | |
CHEBI:35019 | |
266.32 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Suspected of causing cancer.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Obtain special instructions before use.
Wear pro
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-800-2
missing translation for 'rtecsNumber' : TC7700000
missing translation for 'tsca' : TSCA
- Tributyl phosphate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H351-Suspected of causing cancer.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only