missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >300.0°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 0.97 |
Packaging | Plastic bottle |
Linear Formula | CCl3CO2Na |
Quantity | 100 g |
Fieser | 01,1107; 02,388; 06,553; 07,341 |
Merck Index | 15, 9792 |
Solubility Information | Solubility in water: soluble (153 g/100mL at 20°C). Other solubilities: soluble in ethanol, acetone, diethyl ether |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0459806 | Certificate of Analysis | 02/02/2024 | 184150025,184155000,184151000 |
A0452037 | Certificate of Analysis | 02/05/2023 | 184150025,184155000,184151000 |
A0394440 | Certificate of Analysis | 15/12/2022 | 184150025,184155000,184151000 |
A0408741 | Certificate of Analysis | 13/12/2022 | 184150025,184155000,184151000 |
A0411461 | Certificate of Analysis | 13/12/2022 | 184150025,184155000,184151000 |
Safety and Handling


Product Identifier
- Trichloroacetic acid, sodium salt
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity Category 3
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Sodium trichloroacetate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Trichloroacetic acid, sodium salt, 97%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction