missing translation for 'onlineSavingsMsg'
Learn More

Tricine, ≥98.5% (Dry Basis), Ultrapure, Thermo Scientific Chemicals

£175.00 - £277.00

Chemical Identifiers

CAS 5704-04-1
Molecular Formula C6H13NO5
Molecular Weight (g/mol) 179.17
MDL Number MFCD00004277
InChI Key SEQKRHFRPICQDD-UHFFFAOYSA-N
Synonym N-Tris(Hydroxymethyl)methylglycine
PubChem CID 79784
ChEBI CHEBI:39063
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
SMILES C(C(=O)O)NC(CO)(CO)CO
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15835478
View Documents
Thermo Scientific
J22561.36
500 g Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15845478
View Documents
Thermo Scientific
J22561.A1
1 kg Plastic bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

5704-04-1
179.17
SEQKRHFRPICQDD-UHFFFAOYSA-N
79784
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
C6H13NO5
MFCD00004277
N-Tris(Hydroxymethyl)methylglycine
CHEBI:39063
C(C(=O)O)NC(CO)(CO)CO

Specifications

5704-04-1
Crystalline Powder
(CH2OH)3CNHCH2COOH
1937804
SEQKRHFRPICQDD-UHFFFAOYSA-N
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]acetic acid
79784
179.17
Dry basis
∼183°C (decomposition)
C6H13NO5
MFCD00004277
N-Tris(Hydroxymethyl)methylglycine
C(C(=O)O)NC(CO)(CO)CO
179.17
CHEBI:39063
≥98.5%
Tricine
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only