Learn More
Triethanolamine, 99+%, Thermo Scientific Chemicals
£724.00
Chemical Identifiers
CAS | 102-71-6 |
---|---|
Molecular Weight (g/mol) | 149.19 |
MDL Number | MFCD00002855 |
InChI Key | GSEJCLTVZPLZKY-UHFFFAOYSA-N |
Synonym | triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35 |
PubChem CID | 7618 |
ChEBI | CHEBI:28621 |
IUPAC Name | 2-[bis(2-hydroxyethyl)amino]ethanol |
SMILES | C(CO)N(CCO)CCO |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
102-71-6 | |
MFCD00002855 | |
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35 | |
CHEBI:28621 | |
C(CO)N(CCO)CCO |
149.19 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
7618 | |
2-[bis(2-hydroxyethyl)amino]ethanol |
Specifications
102-71-6 | |
1.1245g/mL | |
99+% | |
(HOCH2CH2)3N | |
MFCD00002855 | |
15, 9830 | |
GSEJCLTVZPLZKY-UHFFFAOYSA-N | |
2-[bis(2-hydroxyethyl)amino]ethanol | |
7618 | |
149.19 | |
Oily Liquid |
Colorless to Yellow | |
Authentic | |
C6H15NO3 | |
1.4830 to 1.4860 (20°C, 589nm) | |
01,1196 | |
triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35 | |
C(CO)N(CCO)CCO | |
149.19 | |
CHEBI:28621 | |
≥99.0% | |
Triethanolamine |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only