Learn More
Triethylenetetramine, 60%, Thermo Scientific Chemicals
£20.60 - £109.00
Chemical Identifiers
CAS | 112-24-3 |
---|---|
Molecular Formula | C6H18N4 |
Molecular Weight (g/mol) | 146.24 |
MDL Number | MFCD00008169 |
InChI Key | VILCJCGEZXAXTO-UHFFFAOYSA-N |
Synonym | triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 |
PubChem CID | 5565 |
ChEBI | CHEBI:39501 |
IUPAC Name | N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
SMILES | C(CNCCNCCN)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10633522
|
Thermo Scientific Acros
344070500 |
50 g |
£20.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10603712
|
Thermo Scientific Acros
344075000 |
500 g |
£36.60
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15294704
|
Thermo Scientific Acros
344070025 |
2.5 kg |
£109.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-24-3 | |
146.24 | |
VILCJCGEZXAXTO-UHFFFAOYSA-N | |
5565 | |
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine |
C6H18N4 | |
MFCD00008169 | |
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 | |
CHEBI:39501 | |
C(CNCCNCCN)N |
Specifications
112-24-3 | |
0.9800g/mL | |
129°C | |
55% min. (GC) | |
C6H18N4 | |
1.4960 to 1.5000 | |
04,255 | |
15,9828 | |
triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951 | |
VILCJCGEZXAXTO-UHFFFAOYSA-N | |
N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine | |
5565 | |
146.24 | |
Technical | |
Triethylenetetramine, 60% |
-35.0°C | |
280.0°C | |
Authentic | |
Glass bottle | |
H2NCH2CH2NHCH2CH2NHCH2CH2NH2 | |
MFCD00008169 | |
01,1204 | |
0.98 | |
Solubility in water: soluble in water. Other solubilities: soluble in alcohol | |
C(CNCCNCCN)N | |
146.24 | |
CHEBI:39501 | |
60% | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful in contact with skin.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-950-6
missing translation for 'rtecsNumber' : YE6650000
missing translation for 'tsca' : TSCA
- Triethylenetetramine
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only