missing translation for 'onlineSavingsMsg'
Learn More
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™ Product Code.: 10294110

Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™

Product Code. 10294110
100 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 10294110

Brand: Fisher Chemical T/3258/PB05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C2HF3O2, CAS Number-76-05-1, acetic acid, trifluoro, cf3cooh, kyselina trifluoroctova, perfluoroacetic acid, trifluoroacetic acid, trifluoroaceticacid, trifluoro acetic acid, trifluoro-acetic acid, trifluoroacetic acid, trifluoroethanoic acid, 100mL, 72 deg.C, CHEBI:45892, Colorless, 114.02g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctovaShow MoreShow Less
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

pH 0.45
Melting Point -15°C
Boiling Point 72°C
Color Colorless
IUPAC Name 2,2,2-trifluoroacetic acid
Formula Weight 114.02g/mol
Quantity 100 mL
Packaging Plastic Coated Glass bottle
Physical Form Liquid
Vapor Density 3.9
Vapor Pressure 107mbar 25
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
  • P273-Avoid release to the environment.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trifluoroacetic Acid, 99+%, for HPLC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.