missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, For protein sequencing, Honeywell Fluka™
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Linear Formula | CF3COOH |
UN Number | UN2699 |
Formula Weight | 114.02 |
Percent Purity | ≥99% |
Density | 1.489g/mL at 20°C |
Quantity | 100 g |
Flash Point | >100°C (>212°F) |
Vapor Density | 3.9 (vs air) |
Vapor Pressure | 97.5mmHg (20°C) |
Chemical Name or Material | Trifluoroacetic Acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'