missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic Acid, HPLC Grade, J.T. Baker™
3405.75 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
70 mL
900 mL
Packaging:
Can
Narrow Mouth Amber Glass Bottle
Specifications
Formula Weight | 114.03 |
Grade | HPLC |
Quantity | 900 mL |
Packaging | Narrow Mouth Amber Glass Bottle |
Specific Gravity | 1L = 1.53kg |
Chemical Name or Material | Trifluoroacetic Acid, Gradient Tested for HPLC Peptide and Protein Analysis |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS05_Corrosive.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- trifluoroacetic acid
- Danger
- Skin corrosion/irritation Category 1 A
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'