missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic Acid, LC-MS Ultra, Honeywell™ Fluka™

Product Code. 15668310
Click to view available options
Quantity:
1 mL
2 mL
50 mL
Packaging:
Ampoule
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

LC-MS Ultra, Eluent additive for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15°C
Boiling Point 72°C
Color Colorless
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
Solubility Information Soluble in water
IUPAC Name trifluoroacetic acid
Formula Weight 114.02g/mol
Grade LC-MS Ultra
Density 1.489g/mL at 20°C
Quantity 2 mL
Flash Point >100°C (>212°F)
Packaging Ampoule
Physical Form Liquid
Refractive Index n20/D 1.3(lit.)
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5mmHg (20°C)
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'