missing translation for 'onlineSavingsMsg'
Learn More
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™ Product Code.: 10125637

Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™

Product Code. 10125637
10 mL, Ampules
Click to view available options
Quantity:
0.5 mL
1 mL
2 mL
10 mL
50 mL
Packaging:
Amber Glass
Ampule
Ampules
This item is not returnable. View return policy

Product Code. 10125637

Brand: Fisher Chemical A11610X1AMP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

Trifluoroacetic Acid, >-99.5%, C2HF3O2, CAS Number-76-05-1, trifluoroethanoic acid, trifluoroacetic acid, cf3cooh, acetic acid, trifluoro, trifluoro-acetic acid, perfluoroacetic acid, kyselina trifluoroctova, trifluoro acetic acid, trifluoroacetic acid, trifluoroaceticacid, 50mL

  • Ideal high purity mobile phase additive for LC/MS and UHPLC-UV applications
  • Evaluated for metal impurities at ppb concentrations for minimal metal mass adduct formation
  • Screened for UV-absorbing contaminants to reduce interferences and for smoother baselines
  • TFA is a common mobile phase additive that can function as an ion pair reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctovaShow MoreShow Less
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Absorbance 0.003 AU max. at 254nm, 0.03 AU max. at 300nm, 0.06 AU max. at 230nm, 0.06 AU max. at 275nm, 0.18 AU max. at 220nm, 0.55 AU max. at 210nm
pH 0.45
Melting Point -15°C
Boiling Point 72°C
Color Colorless
Assay Percent Range ≥99.5 %
Linear Formula CF3COOH
Merck Index 15, 9847
Formula Weight 114.02
Percent Purity ≥99.5%
Grade OPTIMA™ LC-MS
Density 1.535g/cm³
Quantity 10 mL
Packaging Ampules
Physical Form Liquid
Residue after Evaporation 1ppm max.
Vapor Density 3.9
Vapor Pressure 107mbar 25
Chemical Name or Material Trifluoroacetic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • TRIFLUOROACETIC ACID
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Trifluoroacetic Acid, Optima™ LC/MS Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.