missing translation for 'onlineSavingsMsg'
Learn More

Trifluoroacetic acid, Reagent Grade, 99%, Honeywell™

Product Code. 15618200
Click to view available options
Quantity:
1 Ampule
1 L
10 Ampules
10 x 10 Ampules
100 mL
2 L
25 mL
5 mL
5 x 10 Ampules
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-05-1
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 114.02
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
Synonym trifluoroacetic acid, perfluoroacetic acid, trifluoroethanoic acid, trifluoroacetic acid, acetic acid, trifluoro, trifluoroaceticacid, cf3cooh, trifluoro acetic acid, trifluoro-acetic acid, kyselina trifluoroctova
PubChem CID 6422
ChEBI CHEBI:45892
IUPAC Name trifluoroacetic acid
SMILES OC(=O)C(F)(F)F

Specifications

Melting Point -15.4°C (literature)
Boiling Point 72°C
CAS Max % 100.0000%
Linear Formula CF3COOH
UN Number UN2699
Beilstein 742035
Formula Weight 114.02
Percent Purity ≥99%
Grade Reagent
Density 1.489 g/mL at 20 °C(lit.)
Quantity 2 L
Flash Point 100°C
Packaging Glass Bottle
Vapor Density 3.9 (vs air)
Vapor Pressure 97.5 mmHg (20°C)
Chemical Name or Material Trifluoroacetic acid, Reagent Grade, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Trifluoroacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 A
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.