missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trifluoroacetic acid, Reagent Grade, 99%, Honeywell™
Click to view available options
Quantity:
1 Ampule
1 L
10 Ampules
10 x 10 Ampules
100 mL
2 L
25 mL
5 mL
5 x 10 Ampules
500 mL
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Linear Formula | CF3COOH |
UN Number | UN2699 |
Beilstein | 742035 |
Formula Weight | 114.02g/mol |
Percent Purity | 99% |
Grade | ReagentPlus™ |
Density | 1.489g/mL at 20°C |
Quantity | 100 mL |
Flash Point | >100°C (>212°F) |
Vapor Density | 3.9 (vs air) |
Show More |
Safety and Handling
Product Identifier
- Trifluoroacetic acid
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction