Learn More
Trimethylacetyl chloride, 99%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -56.0°C |
Color | Colorless to Pink |
Density | 0.9800g/mL |
Boiling Point | 105.0°C |
Flash Point | 14°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Linear Formula | (CH3)3CCOCl |
Refractive Index | 1.4110 to 1.4130 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0445409 | Certificate of Analysis | 26/02/2024 | 140050050,140052500,140050010,140051000 |
A0436965 | Certificate of Analysis | 25/08/2023 | 140050050,140052500,140050010,140051000 |
A0445616 | Certificate of Analysis | 05/09/2022 | 140050050,140052500,140050010,140051000 |
A0444664 | Certificate of Analysis | 01/08/2022 | 140050050,140052500,140050010,140051000 |
A0422560 | Certificate of Analysis | 14/03/2022 | 140050050,140052500,140050010,140051000 |
Safety and Handling



- Trimethylacetyl chloride
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H302-Harmful if swallowed.
- H290-May be corrosive to metals.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.