missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution, Thermo Scientific Chemicals
£22.70 - £270.00
Specifications
Chemical Name or Material | Trimethylamine |
---|---|
Color | Colorless |
CAS | 75-50-3, 7732-18-5 |
Boiling Point | 30.0°C to 100.0°C |
Melting Point | -2.0°C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10623003
|
Thermo Scientific Acros
370785000 |
500 mL | Glass bottle |
£22.70
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10364262
|
Thermo Scientific Acros
370780010 |
1 L | Glass bottle |
£37.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10120812
|
Thermo Scientific Acros
370780025 |
2.5 L | Glass bottle |
£72.10
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11965161
|
Thermo Scientific Acros
370780100 |
10 L | Plastic drum |
£270.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Trimethylamine | |
75-50-3, 7732-18-5 | |
-2.0°C | |
(CH3)3N | |
0.8600g/mL | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, benzene, toluene,, xylene, ethylbenzene and chloroform | |
CN(C)C | |
59.11 | |
CHEBI:18139 | |
48 to 52% (total base) | |
0.86 | |
Liquid |
Colorless | |
30.0°C to 100.0°C | |
C3H9N | |
MFCD00008327 | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
N,N-dimethylmethanamine | |
1146 | |
59.11 | |
Pure | |
−45°C |
Safety and Handling
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Trimethylamine, 45-50 wt.% aqueous solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H224-Extremely flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only