Learn More
Trimethylamine, pure, 7.3M (50 wt.%) aqueous solution, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 370780100
281.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Trimethylamine | |
75-50-3, 7732-18-5 | |
-2.0°C | |
(CH3)3N | |
0.8600g/mL | |
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether, benzene, toluene,, xylene, ethylbenzene and chloroform | |
CN(C)C | |
59.11 | |
CHEBI:18139 | |
48 to 52% (total base) | |
Plastic drum | |
10 L | |
Liquid |
Colorless | |
30.0°C to 100.0°C | |
C3H9N | |
MFCD00008327 | |
trimethylamine, methanamine, n,n-dimethyl, dimethylmethaneamine, n-trimethylamine, trimethylamine solution, ch3 3n, trimethyl amine, trimethylamin, trimethyl-amine, fema number 3241 | |
GETQZCLCWQTVFV-UHFFFAOYSA-N | |
N,N-dimethylmethanamine | |
1146 | |
59.11 | |
Pure | |
0.86 | |
−45°C |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Trimethylamine, 45-50 wt.% aqueous solution
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 1
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H224-Extremely flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0453323 | Certificate of Analysis | 25/08/2023 | 370780025,370780010,370780100,370785000 |
A0401878 | Certificate of Analysis | 27/02/2023 | |
A0401878 | Certificate of Analysis | 14/12/2022 | 370780025,370780010,370780100,370785000 |
A0408791 | Certificate of Analysis | 13/12/2022 | 370780025,370780010,370780100,370785000 |
A0413571 | Certificate of Analysis | 13/12/2022 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only