Learn More
(Trimethylsilyl)methyllithium, 0.7M (10 wt%) solution in hexanes, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 377451000
97.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagentSpecifications
(Trimethylsilyl)methyllithium | |
1822-00-0, 92112-69-1, 92112-69-1 | |
14.0, 93.0 | |
0.6500g/mL | |
−40°C | |
C4H11LiSi | |
MFCD00010747 | |
0.65 | |
KVWLUDFGXDFFON-UHFFFAOYSA-N | |
lithium;methanidyl(trimethyl)silane | |
3482579 | |
7 to 14 wt% |
0.8M (12 wt%) solution in hexanes | |
7.0, 86.0 | |
Brown-Yellow to Colorless | |
65.0°C to 70.0°C | |
AcroSeal™ Glass bottle | |
LiCH2Si(CH3)3 | |
100 mL | |
trimethylsilyl methyllithium, trimethylsilyl methyl lithium, lich2sime3, pubchem18144, tms-ch2li, lithiomethyltrimethylsilane, acmc-1bvf0, trimethylsilylmethyl lithium | |
[Li+].C[Si](C)(C)[CH2-] | |
94.16 | |
94.16 | |
Solution |
Safety and Handling
- (Trimethylsilyl)methyllithium, 0.7M (10wt%) solution in hexanes
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H314-Causes severe skin burns and eye damage.
- H336-May cause drowsiness or dizziness.
- H361f-Suspected of damaging fertility.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- EUH014-Reacts violently with water.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only