Learn More
Trimethylsilyl trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
£33.30 - £413.00
Chemical Identifiers
CAS | 27607-77-8 |
---|---|
Molecular Formula | C4H9F3O3SSi |
Molecular Weight (g/mol) | 222.26 |
MDL Number | MFCD00000406 |
InChI Key | FTVLMFQEYACZNP-UHFFFAOYSA-N |
Synonym | trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate |
PubChem CID | 65367 |
IUPAC Name | trimethylsilyl trifluoromethanesulfonate |
SMILES | C[Si](C)(C)OS(=O)(=O)C(F)(F)F |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10042630
|
Thermo Scientific Acros
209440100 |
10 mL | Glass bottle |
£33.30
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042640
|
Thermo Scientific Acros
209440500 |
50 mL | Glass bottle |
£102.00
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10237473
|
Thermo Scientific Acros
209442500 |
250 mL | Glass bottle |
£413.00
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
27607-77-8 | |
222.26 | |
FTVLMFQEYACZNP-UHFFFAOYSA-N | |
65367 | |
C[Si](C)(C)OS(=O)(=O)C(F)(F)F |
C4H9F3O3SSi | |
MFCD00000406 | |
trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate | |
trimethylsilyl trifluoromethanesulfonate |
Specifications
27607-77-8 | |
1.2250g/mL | |
25°C | |
99% | |
CF3SO3Si(CH3)3 | |
MFCD00000406 | |
15, 9891 | |
trimethylsilyl triflate, tmsotf, trifluoromethanesulfonic acid trimethylsilyl ester, trimethylsilyl trifluoromethylsulphonate, unii-z84v0cbh9j, methanesulfonic acid, trifluoro-, trimethylsilyl ester, trimethylsilyl trifluoromethylsulfonate, trimethylsllytrifluoromethanesulphonate, trimethylsilyl trifluoromethanesulphonate, trimethylsilyltrifluoromethanesulphonate | |
FTVLMFQEYACZNP-UHFFFAOYSA-N | |
trimethylsilyl trifluoromethanesulfonate | |
65367 | |
99% | |
Trimethylsilyl trifluoromethanesulfonate |
Colorless to Brown | |
140°C | |
Authentic | |
C4H9F3O3SSi | |
1.3600 to 1.362 | |
08,514; 09,497; 10,438; 11,584; 12,543; 13,118; 14,333; 16,363; 17,379 | |
1.225 | |
Solubility in water: reacts | |
C[Si](C)(C)OS(=O)(=O)C(F)(F)F | |
222.26 | |
222.26 | |
Fuming Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 248-565-4
- Trimethylsilyl trifluoromethanesulfonate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only