missing translation for 'onlineSavingsMsg'
Learn More
Triphenylphosphine, 99%, Thermo Scientific Chemicals Product Code.: 10337120

Triphenylphosphine, 99%, Thermo Scientific Chemicals

Product Code. 10337120
250 g, Plastic bottle
Click to view available options
Quantity:
25 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10337120

Brand: Thermo Scientific Acros 140422500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 603-35-0
Molecular Formula C18H15P
Molecular Weight (g/mol) 262.29
MDL Number MFCD00003043 MFCD20489348
InChI Key RIOQSEWOXXDEQQ-UHFFFAOYSA-N
Synonym triphenylphosphine, triphenyl phosphine, phosphine, triphenyl, triphenylphosphorus, triphenyl-phosphane, triphenylphosphide, phosphorustriphenyl, trifenylfosfin, trifenylfosfin czech, triphenylphosphine resinShow MoreShow Less
PubChem CID 11776
IUPAC Name triphenylphosphane
SMILES C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 78.5°C to 81.5°C
Color White
Boiling Point 377.0°C
Flash Point 181°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5)3P
Quantity 250 g
Beilstein 16,759
Fieser 01,1238; 02,443; 03,317; 04,548; 05,725; 06,643; 07,403; 11,588; 12,550; 13,308; 14,60; 15,248; 16,264; 17,389
Merck Index 15,9916
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in glacial acetic acid,, soluble in benzene, chloroform, acetone and ccl4,, freely soluble in diethyl ether,, slightly soluble in alcohol
Formula Weight 262.28
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material Triphenylphosphine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467618 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number,140422500,140420250,140420010
Lot NumberA0466083 Certificate TypeCertificate of Analysis Date19/09/2024 Catalog Number140420050,140422500,140420250,140420010
Lot NumberA0460540 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number140420050,140422500,140420250,140420010
Lot NumberA0457490 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number140420050,140422500,140420250,140420010
Lot NumberA0449436 Certificate TypeCertificate of Analysis Date17/01/2023 Catalog Number140420050,140422500,140420250,140420010
compliance-icons compliance-icons
Product Identifier
  • Triphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Triphenylphosphine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.