missing translation for 'onlineSavingsMsg'
Learn More

Triphenyltin chloride, 95%, Thermo Scientific Chemicals

£21.41 - £165.83

Chemical Identifiers

CAS 639-58-7
Molecular Formula C18H17ClSn
Molecular Weight (g/mol) 387.49
MDL Number MFCD00000519
InChI Key ZDYYCRIXXMWWQC-UHFFFAOYSA-N
Synonym triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
PubChem CID 12540
ChEBI CHEBI:35036
IUPAC Name chloro(triphenyl)stannane
SMILES Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11359085
View Documents
Thermo Scientific Alfa Aesar
L14289.14
25 g
£21.41
25g
Please to purchase this item. Need a web account? Register with us today!
11369085
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L14289.22
100 g
£43.22
100g
Please to purchase this item. Need a web account? Register with us today!
11379085
View Documents
Thermo Scientific Alfa Aesar
L14289.36
500 g
£165.83
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triphenyltin chloride is an important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water (0.005g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is hygroscopic in nature and sensitive to moisture.
Specifications

Chemical Identifiers

639-58-7
387.49
ZDYYCRIXXMWWQC-UHFFFAOYSA-N
12540
chloro(triphenyl)stannane
C18H17ClSn
MFCD00000519
triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
CHEBI:35036
Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1

Specifications

639-58-7
240°C (13.5 mmHg)
Unpleasant
MFCD00000519
524762
triphenyltin chloride, fentin chloride, chlorotriphenyltin, triphenylchlorotin, stannane, chlorotriphenyl, brestanol, tptc, triphenylchlorostannane, aquatin, tinmate
ZDYYCRIXXMWWQC-UHFFFAOYSA-N
chloro(triphenyl)stannane
12540
385.48
Triphenyltin chloride
103°C to 106°C
>110°C (230°F)
C18H17ClSn
UN3146
Moisture sensitive
Soluble in water (0.005g/L at 20°C).
Cl.C1=CC=C(C=C1)[SnH](C1=CC=CC=C1)C1=CC=CC=C1
387.49
CHEBI:35036
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

missing translation for 'einecsNumber' : 211-358-4

missing translation for 'rtecsNumber' : WH6860000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only