Learn More
BTX™ Triple Electroporation Tweezertrode
Triple Electroporation Tweezertrode is a reusable non-invasive, tweezer-style electrode for drug or gene delivery in animal tissues.
Brand:  BTX™ 450494
 View more versions of this product
Product Code. 15625028
  • £4750.00 / Each
Explore more special offers
Description
Description
Triple Tweezertrodes have:
  • Three electrode contacts with adjustable position and polarities
  • Improves electrical field distribution for in utero electroporation applications
Compatability:
  • ECM830
  • ECM2001+
  • Gemini X2
  • Connect to BTX generators using the Tweezertrode Connection cables (45-0204)
Applications:
  • In vivo drug or gene delivery
  • Ex vivo drug or gene delivery
  • In utero drug or gene delivery
Tweezertrodes may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
Specifications
Specifications
Not Autoclavable
Reusable
Non-sterile
0.2 in.
Tweezers
ECM 830, ECM 2001, Gemini X2
Cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
5 mm