Learn More
BTX™ Triple Electroporation Tweezertrode
Triple Electroporation Tweezertrode is a reusable non-invasive, tweezer-style electrode for drug or gene delivery in animal tissues.
Brand:  BTX™ 450493
 View more versions of this product
Product Code. 15741078
  • £4555.00 / Each
Estimated Shipment: 03-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Triple Tweezertrodes have:
  • Three electrode contacts with adjustable position and polarities
  • Improves electrical field distribution for in utero electroporation applications
Compatability:
  • ECM830
  • ECM2001+
  • Gemini X2
  • Connect to BTX generators using the Tweezertrode Connection cables (45-0204)
Applications:
  • In vivo drug or gene delivery
  • Ex vivo drug or gene delivery
  • In utero drug or gene delivery
Tweezertrodes may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
Specifications
Specifications
Not Autoclavable
Reusable
Non-sterile
0.12 in.
0 to 200 VDC (Do Not Use AC Current)
Tweezers
ECM 830, ECM 2001, Gemini X2
5°C to 40°C
3 mm