missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One Triple-Packed Serological Pipette
Packed in three individual peel bags for sterile transfer of products.
Brand: Greiner Bio-One 607160-TRI
This item is not returnable.
View return policy
Description
- Made from polystyrene
- Free of heavy metal
- Not autoclavable
- Clear graduated pipette
- Sterile
Specifications
± 0.2% | |
Polystyrene | |
Yes | |
Triple packed | |
10 mL |
Includes yellow code ring, cotton plug | |
Sterile | |
1/10 mL | |
Clear | |
Pipette |