missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine Product Code.: 15476030
  • ONLINE EXCLUSIVE PRICE
    ÂŁ535.00
    ÂŁ300.21 / Pack of 100
    Save ÂŁ234.79
    43% Off

Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine

Product Code. 15476030
100/Pk., 55mm Contact Plate, Tryptone Soya Agar, Germany, Pack of 100
Click to view available options
Quantity:
100/Pk.
Format:
55mm Contact Plate
90mm Monoplate
Media Type:
Tryptone Soya Agar
Manufacturing Origin:
Germany
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15476030

Brand: Thermo Scientific™ PO5511D

  • ONLINE EXCLUSIVE PRICE
    ÂŁ535.00
    ÂŁ300.21 / Pack of 100
    Save ÂŁ234.79
    43% Off
View Available Promotions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates for passive and active air sampling applications.

Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.

Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.

Three protective layers reduce risk of contamination:
  • Primary film with moisture control patch
  • Secondary film VHP barrier with immobilized desiccant sachet
  • Tertiary film dust cover

  • The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
     
  • Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs

Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY

Specifications

Color Ivory, transparent
Content And Storage 2 to 25°C
Format 55mm Contact Plate
Manufacturing Origin Germany
Media Type Tryptone Soya Agar
Product Type Agar
Fill Weight 13.0g ± 0.5g
Packaging Type Triple Wrapped - Irradiated
Quantity 100/Pk.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number6333185 Certificate TypeCertificate of Analysis Date03/11/2025 Product Code
Lot Number6333184 Certificate TypeCertificate of Analysis Date03/11/2025 Product Code
Lot Number6330121 Certificate TypeCertificate of Analysis Date28/10/2025 Product CodePO5511D
Lot Number6330122 Certificate TypeCertificate of Analysis Date28/10/2025 Product CodePO5511D
Lot Number6327319 Certificate TypeCertificate of Analysis Date21/10/2025 Product CodePO5511D
Promo
Expires: 2025-12-31
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.

Applying Promo and Adding...

Promo
Expires: 2025-11-07
Microbiology Madness
Price slashed on over 3000 products

Applying Promo and Adding...

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.