Learn More
Thermo Scientific™ Pro-Tect Agar TWI Contact Plate (TSA Agar with inhibitors)
Effortlessly sample surfaces after cleaning with triple wrapped, irradiated plates with disinhibitors to neutralize surface disinfectants and aid in microorganism recovery.
Brand: Thermo Scientific™ PO0678D
33.62 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Description
Effortlessly sample surfaces after cleaning and disinfection using Thermo Scientific™ Pro-Tect Agar TWI Contact Plates, containing Lecithin, polysorbate 80, sodium thiosulfate and Tween 80 to neutralize surface disinfectants and aid in microorganism recovery. The plates are triple-wrapped and irradiated for sterility.
Streamline environmental monitoring of cleanroom environments with Pro-Tect Agar triple-wrapped, irradiated contact plates for surface sampling.- Secure: dome-shaped bottom prevents agar from falling out of the dish
- Easy-to-handle: GripFit™ technology ensures the lid remains in place during handling while being easily removed during environmental monitoring
- Straightforward results interpretation: inner layer on the plate base is sub-divided into grids to allow easy calculation of colony forming units (CFU) from surface tests, enabling labs to report whether contaminant levels fall within pre-defined acceptable limits
- Easy-to-stack: provides more stability during handling and transport, and ventilation is reduced to prevent plates from drying out
- Consistent: manufactured using a specialized, automated procedure to ensure a smooth, evenly distributed layer of agar on each plate
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
2°C to 25°C | |
90mm Monoplate | |
13 mL |
Pro-Tect TWI | |
Agar | |
1 x 20 Plates |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6239762 | Certificate of Analysis | 11/03/2025 | PO0678D |
6234192 | Certificate of Analysis | 24/02/2025 | PO0678D |
6230541 | Certificate of Analysis | 18/02/2025 | PO0678D |
6228475 | Certificate of Analysis | 04/02/2025 | PO0678D |
6228476 | Certificate of Analysis | 04/02/2025 | PO0678D |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.