missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine
Carry out environmental monitoring in your most regulated environments using Thermo Scientific™ Triple Wrapped Plates for passive and active air sampling applications.
£240.00 - £451.00
Specifications
Color | Ivory, transparent |
---|---|
Content And Storage | 2 to 25°C |
Description | Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine |
Manufacturing Origin | Germany |
Product Type | Agar |
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Manufacturing Origin | Price | Quantity & Availability | |||
15436030
|
Thermo Scientific™
PO5501B |
Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine | 100/Pk. | Germany |
£240.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
15476030
|
Thermo Scientific™
PO5511D |
Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine | 100/Pk. | Germany |
£451.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
Description
Experience next-level cleanroom confidence with Thermo Scientific™ Triple Wrap Sterile Pack Tryptone Soya Agar plates, complete with VHP indicator, Lecithin, Polysorbate 80, Sodium Thiosulphate and L-Histidine.
Achieve confidence in your results and ensure optimum media fertility using plates with a Vapour-phase Hydrogen Peroxide (VHP) exposure indicator, designed to confirm the integrity of the wrapping material and seal during VHP exposure.Three protective layers reduce risk of contamination:
- Primary film with moisture control patch
- Secondary film VHP barrier with immobilized desiccant sachet
- Tertiary film dust cover
The incorporation of novel elements enhance moisture control by permitting the passage and absorption of excess moisture
Using plates with an improved shelf life and room temperature storage offers a cost-effective solution, minimizing new batch validation and refrigeration costs
Not all products are available for sale in all territories. Please inquire.
Specifications
Ivory, transparent | |
Triple Wrap Sterile Pack w/VHP Indicator Tryptone Soya Agar w/Lecithin, Polysorbate 80, Sodium Thiosulphate, L-Histidine | |
Agar | |
100/Pk. |
2 to 25°C | |
Germany | |
Triple Wrapped - Irradiated |