missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIS, 0.5M buffer soln., pH 7.4, Thermo Scientific™
£54.80 - £69.10
Chemical Identifiers
CAS | 77-86-1 |
---|---|
Molecular Formula | C4H11NO3 |
Molecular Weight (g/mol) | 121.136 |
MDL Number | MFCD00004679 |
InChI Key | LENZDBCJOHFCAS-UHFFFAOYSA-N |
PubChem CID | 6503 |
ChEBI | CHEBI:9754 |
IUPAC Name | 2-amino-2-(hydroxymethyl)propane-1,3-diol |
SMILES | C(C(CO)(CO)N)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15455079
|
Thermo Scientific Alfa Aesar
J62778.AP |
500 mL |
£54.80
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15465079
|
Thermo Scientific Alfa Aesar
J62778.K2 |
1 L |
£69.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesStore away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
77-86-1 | |
121.136 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
CHEBI:9754 | |
C(C(CO)(CO)N)O |
C4H11NO3 | |
MFCD00004679 | |
6503 | |
2-amino-2-(hydroxymethyl)propane-1,3-diol |
Specifications
77-86-1 | |
7.4 | |
C4H11NO3 | |
Not miscible or difficult to mix in water. Soluble in ethylene glycol (79.1mg/ml),methanol (26mg/ml),anhydrous ethanol (14.6mg/ml),95% ethanol (22mg/ml),DMF (14mg/ml),acetone (2mg/ml),ethyl acetate (0.5mg/ml),olive oil (0.4mg/ml),chloroform (0.05mg/ml),and alkaline solution. | |
C(C(CO)(CO)N)O | |
121.136 | |
CHEBI:9754 |
0.5 M | |
Liquid | |
MFCD00004679 | |
LENZDBCJOHFCAS-UHFFFAOYSA-N | |
2-amino-2-(hydroxymethyl)propane-1,3-diol | |
6503 | |
TRIS buffer soln. |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures